Learn More
Epredia™ Lab Vision™ Rhodopsin (Opsin) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: RET-P1
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Membrane preparation from adult rat retina
Molecular Weight: 39kDa
Positive Control: Brain or retina
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Rhodopsin (Opsin) Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | RET-P1 |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Membrane preparation from adult rat retina |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.