Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100 Protein Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS296RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS296RQ 12 mL Unconjugated Ready-to-Use Prediluted
MS296PABX 200 μL Unconjugated Concentrated 1mg/mL
MS296P1 500 μL Unconjugated Concentrated 200μg/mL
MS296B0 100 μL Biotin Concentrated 200μg/mL
MS296B1 500 μL Biotin Concentrated 200μg/mL
MS296B 1 mL Biotin Concentrated 200μg/mL
MS296P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS296P 1 mL Unconjugated Concentrated 200μg/mL
MS296R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS296RQ Supplier Epredia Supplier No. MS296RQ
Add to Cart
Add to Cart

Provide accurate, reproducible results with the Epredia™ S100 Protein Ab-1, Mouse Monoclonal Antibody..

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Mouse

Clone: 4C4.9

Isotype: IgG2a

Species Reactivity: Human, Cow, Rat and Mouse. Others not known.

Epitope: Not determined

Immunogen: Purified bovine brain S100 protein

Molecular Weight: ∽22kDa (nonreduced); ∽11kDa (reduced)

Positive Control: Melanoma or Schwannoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen S100 Protein Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4C4.9
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified bovine brain S100 protein
Quantity 12 mL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.