Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ S100 Protein Ab-2 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB044A1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB044A1 500 μL Concentrated
RB044R1 1 mL Prediluted
RB044R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RB044A1 Supplier Epredia™ Supplier No. RB044A1

Epredia™ S100 Protein Ab-2, Rabbit Polyclonal Antibody provides accurate, reproducible results.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. Antibody to S100 stains Schwannomas, ependymomas, astrogliomas, almost all benign and malignant melanomas and their metastases. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Cow, Kangaroo, Mouse, Rat and Chicken. Others not tested.

Epitope: Not determined

Immunogen: S100 protein purified from cow brain

Molecular Weight: ∽22kDa (nonreduced); ∽11kDa (reduced)

Positive Control: Melanoma or Schwannoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunocytochemistry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen S100 Protein Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen S100 protein purified from cow brain
Purification Method Melanoma or Schwannoma
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Kangaroo, Mouse
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.