Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ S100 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB9018P Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9018P 1 mL Concentrated
RB9018R1 1 mL Prediluted
RB9018RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RB9018P Supplier Epredia™ Supplier No. RB9018P

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ S100, Rabbit Polyclonal Antibody.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. Antibody to S100 stains Schwannomas, ependymomas, astrogliomas, almost all benign and malignant melanomas and their metastases. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant protein encoding human S100 protein

Molecular Weight: 21-24kDa

Positive Control: Melanoma or Schwannoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen S100
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein encoding human S100 protein
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.