Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Serotonin Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1431R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1431R1 1 mL Prediluted
MS1431S1 500 μL Concentrated
MS1431S 1 mL Concentrated
MS1431R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1431R1 Supplier Epredia™ Supplier No. MS1431R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Serotonin Ab-1, Mouse Monoclonal Antibody.

Serotonin-positive cells are present in gastric, duodenal, jejunal, ileal, colonic, and appendiceal mucosae. Positive cells are sparse and usually occur in the lower half of the gastric and intestinal mucosae. In the appendix, in addition to the mucosal serotonin-containing cells, there are single or small clusters of serotonin-positive cells in the sub-epithelial region of the lamina propria.

Host Species: Mouse

Clone: 5HT-H209

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: 5-Hydroxytryptamine hydrochloride [3-(2-aminoethyl)-5-hydroxyindole hydrochloride; serotonin hydrochloride]

Positive Control: Appendix

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)


Specifications

Antigen Serotonin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5HT-H209
Conjugate Unconjugated
Host Species Mouse
Immunogen 5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin)
Quantity 1 mL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.