Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ SREBP-1 (Sterol Regulatory Element Binding Protein-1) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1207P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1207P0 100 μL 200μg/mL
MS1207PABX 200 μL 1mg/mL
MS1207P1 500 μL 200μg/mL
MS1207P 1 mL 200μg/mL
MS1207P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1207P0 Supplier Epredia™ Supplier No. MS1207P0

Epredia™ SREBP-1 (Sterol Regulatory Element Binding Protein-1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

SREBP-1 and -2 are transcription factors which participate in the control of cholesterol homeostasis. SREBPs proteins, which are attached to the endoplasmic reticulum and nuclear envelope, are proteolytically cleaved and thus activated in response to conditions of low cellular sterol. Upon activation of SREBP-1 or -2, an ∽480-500 amino acid, N-terminal cleavage fragment of these proteins enters the nucleus and activates transcription of enzymes and other proteins required for cholesterol synthesis. SCA (SREBP-cleavage activity) and caspase-3 proteases cleave SREBPs. SREBP proteins containing point mutations at caspase-3 cleavage sites (Asp460 in SREBP-1 and Asp468 in SREBP-2) do not become cleaved following induction of apoptosis, suggesting that SREBPs may play some role in apoptotic processes.

Host Species: Mouse

Clone: 2A4

Isotype: IgG1/κ

Species Reactivity: Human and Hamster. Others not known.

Epitope: aa 301-407

Immunogen: Recombinant protein encoding N-terminal amino acids 301-407 (the bHLH/leucine zipper domain) of human SREBP-1

Molecular Weight: 125kDa (precursor) and 60-70kDa (cleaved)

Positive Control: HeLa and IMR5 cells

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen SREBP-1 (Sterol Regulatory Element Binding Protein-1) Ab-1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2A4
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding N-terminal amino acids 301-407 (the bHLH/leucine zipper domain) of human SREBP-1
Quantity 100 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Target Species Hamster, Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.