Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surfactant Protein B (Precursor) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS704B0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS704B0 100 μL Biotin Concentrated 200μg/mL
MS704B1 500 μL Biotin Concentrated 200μg/mL
MS704B 1 mL Biotin Concentrated 200μg/mL
MS704P0 100 μL Unconjugated Concentrated 200μg/mL
MS704P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS704PABX 200 μL Unconjugated Concentrated 1mg/mL
MS704P1 500 μL Unconjugated Concentrated 200μg/mL
MS704R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS704B0 Supplier Epredia Supplier No. MS704B0

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Epredia™ Surfactant Protein B (Precursor) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventillation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.

Host Species: Mouse

Clone: SPB01; same as 1-2-A5-9

Isotype: IgG2a/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant prosurfactant protein B

Molecular Weight: 42-46kDa

Positive Control: Lung adenocarcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Surfactant Protein B (Precursor) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPB01; same as 1-2-A5-9
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Recombinant pro-surfactant protein B
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.