Learn More
Epredia™ Lab Vision™ Survivin Rabbit Polyclonal Antibody with BSA and azide
Survivin, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Supplier: Epredia™ RB9245R7
Description
Survivin is a new inhibitor of apoptosis (IAP) protein. It is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. The overexpression of surviving in cancer may overcome this apoptotic checkpoint and favor aberrant progression of transformed cells through mitosis.Host Species: Rabbit
Species Reactivity: Human and Mouse. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from the C-terminus of human survivin
Molecular Weight: 16.5kDa
Positive Control: Normal pancreas (α-cells) or prostate CA
Cellular Localization: Cytoplasmic and Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Survivin | |
Polyclonal | |
Rabbit | |
7 mL | |
Apoptosis | |
Human, Mouse |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
A synthetic peptide derived from the C-terminus of human survivin | |
RUO | |
Primary | |
Prediluted |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.