Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Ab-4 Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB1461R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
Prediluted
Formulation:
Purified w/o BSA and Azide
Purified with BSA and Azide
Ready-to-use for IHC (UVLP)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Formulation
RB1461R1 1 mL Prediluted Ready-to-use for IHC (UVLP)
RB1461P1 500 μL Concentrated Purified with BSA and Azide
RB1461P1ABX 500 μL Concentrated Purified w/o BSA and Azide
RB1461PABX 1 mL Concentrated Purified w/o BSA and Azide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB1461R1 Supplier Epredia Supplier No. RB1461R1

Provide accurate, reproducible results Epredia™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human synaptophysin

Molecular Weight: 38kDa

Positive Control: IMR-5 cells, Pancreas or Pheochromocytoma or GI tumor

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Synaptophysin Ab-4
Applications Western Blot
Classification Polyclonal
Clone Polyclonal
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Rabbit
Immunogen A synthetic peptide from C-terminus of human synaptophysin
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.