Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Synaptophysin (Neuroendocrine Marker) Rabbit Monoclonal Antibody

Catalog No. RM9111S Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9111S 1 mL Concentrated
RM9111S1 500 μL Concentrated
RM9111R1 1 mL Prediluted
RM9111R7 7 mL Prediluted
RM9111RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM9111S Supplier Epredia™ Supplier No. RM9111S

Synaptophysin (Neuroendocrine Marker), Rabbit MAb is part of a comprehensive ab portfolio. Studies show rabbit mabs are more sensitive and specific than other available markers.

This antibody recognizes a protein of 38kDa, identified as synaptophysin. It labels normal neuroendocrine cells of human adrenal medulla, carotid body, skin, pituitary gland, thyroid, lung, pancreas, gastrointestinal mucosa, Paneth's cells in the gastrointestinal tract and of gastric parietal cells. Neurons in the brain, spinal cord, and retina are also labeled. In combination with anti-chromogranin A and anti-NSE, Ab to synaptophysin is very useful in the identification of normal neuroendocrine cells and neuroendocrine neoplasms.

Host Species: Rabbit

Clone: SP11

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide of human synaptophysin

Molecular Weight: 38kDa

Positive Control: IMR5 cells, Pancreas or Pheochromocytoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen Synaptophysin (Neuroendocrine Marker)
Applications Western Blot
Classification Monoclonal
Clone SP11
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide of human synaptophysin
Quantity 1 mL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.