Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synuclein Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1572P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1572P0 100 μL Concentrated 200μg/mL
MS1572PABX 200 μL Concentrated 1mg/mL
MS1572P1 500 μL Concentrated 200μg/mL
MS1572P1ABX 100 μL Concentrated 1mg/mL
MS1572R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1572P0 Supplier Epredia Supplier No. MS1572P0

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Provide accurate, reproducible results Epredia™ Synuclein Ab-2, Mouse Monoclonal Antibody.

Synnuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's (PD) and breast cancer. In AD, a peptide derived from α-synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of α synuclein has been linked to many familial cases of PD. In breast cancer increased expression of γ synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.

Host Species: Mouse

Clone: syn211

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant α-synuclein

Molecular Weight: ∽14kDa

Positive Control: Brain

Cellular Localization: Neuronal cytosol and presynaptic terminals

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Synuclein Ab-2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone syn211
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human recombinant alpha-synuclein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.