Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAG-72/CA 72-4TM Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS138P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
1 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS138P0 100 μL Concentrated 200μg/mL
MS138PABX 200 μL Concentrated 1mg/mL
MS138P 1 mL Concentrated 200μg/mL
MS138R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS138P0 Supplier Epredia Supplier No. MS138P0

Provide accurate, reproducible results with the Epredia™ TAG-72/CA 72-4 Ab-1, Mouse Monoclonal Antibody.

Tumor-associated glycoprotein (TAG-72) is an oncofetal antigen with properties of a mucin. About 60% of carcinoma patients express TAG-72 in their sera.

Host Species: Mouse

Clone: B72.3

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Dog, Hamster and Rat. Others not known.

Epitope: Mucin-carried-sialylated-Tn epitope

Immunogen: Membrane-enriched fraction of a human breast carcinoma liver metastasis

Molecular Weight: >1000kDa

Positive Control: Breast or lung CA

Cellular Localization: Cell surface and cytoplasmic. The majority of human adenocarcinomas including colorectal, pancreatic, gastric, ovarian, endometrial, mammary, and non-small cell lung cancer display some cell populations that are positive for Ab-1 staining. Weak or no reactivity has been observed with most cell types of normal adult tissue excepting the secretory endometrium. Ab-1 is reportedly useful in distinguishing pulmonary adenocarcinomas (B72.3+) from pleural mesotheliomas (B72.3-).

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry
  • Radioimaging

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen TAG-72/CA 72-4 Ab-1
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone B72.3
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Membrane-enriched fraction of a human breast carcinoma liver metastasis
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Hamster, Human, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.