Learn More
Description
Host Species: Mouse
Clone: TAU-5
Isotype: IgG1
Species Reactivity: Human, Sheep, Cow, Mouse and Rat. Others not known.
Epitope: Middle of Tau
Immunogen: Purified bovine microtubule-associated proteins
Molecular Weight: 45-68kDa
Positive Control: IMR5 cells, Human T98G glioblastoma cells or Alzheimer's disease brain tissue
Cellular Localization: Cytoplasmic Ab-2 reacts with the non-phosphorylated as well as the phosphorylated forms of tau and its epitope maps in the middle of tau. Ab-2 intensely stains the human neurofibrillary tangles, neuropil threads, and neuritic plaques associated with Alzheimer's disease. Ab-2 shows no cross reaction with other MAPs or tubulin.
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | TAU Ab-2 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | TAU-5 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified bovine microtubule-associated proteins |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.