Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS247PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS247PABX 200 μL Unconjugated 1mg/mL
MS247P 1 mL Unconjugated 200μg/mL
MS247P1ABX 100 μL Unconjugated 1mg/mL
MS247B0 100 μL Biotin 200μg/mL
MS247B1 500 μL Biotin 200μg/mL
MS247B 1 mL Biotin 200μg/mL
MS247P1 500 μL Unconjugated 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS247PABX Supplier Epredia Supplier No. MS247PABX
Add to Cart
Add to Cart

Provide accurate, reproducible results with the Epredia™ Tau Ab-2, Mouse Monoclonal Antibody.

Tau proteins are microtubule-associated proteins (MAPs). Alternate splicing of tau mRNA and differential phosphorylation contribute to the heterogeneity of tau.

Host Species: Mouse

Clone: TAU-5

Isotype: IgG1

Species Reactivity: Human, Sheep, Cow, Mouse and Rat. Others not known.

Epitope: Middle of Tau

Immunogen: Purified bovine microtubule-associated proteins

Molecular Weight: 45-68kDa

Positive Control: IMR5 cells, Human T98G glioblastoma cells or Alzheimer's disease brain tissue

Cellular Localization: Cytoplasmic Ab-2 reacts with the non-phosphorylated as well as the phosphorylated forms of tau and its epitope maps in the middle of tau. Ab-2 intensely stains the human neurofibrillary tangles, neuropil threads, and neuritic plaques associated with Alzheimer's disease. Ab-2 shows no cross reaction with other MAPs or tubulin.

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen TAU Ab-2
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TAU-5
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified bovine microtubule-associated proteins
Quantity 200 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat, Sheep
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.