Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Tau Ab-3 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB1429A1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB1429A1 500 μL Concentrated
RB1429A 1 mL Concentrated
RB1429R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RB1429A1 Supplier Epredia™ Supplier No. RB1429A1

Epredia™ Tau Ab-3, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Tau proteins are microtubule-associated proteins (MAPs). Alternate splicing of tau mRNA and differential phosphorylation contribute to the heterogeneity of tau.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: Recombinant protein encoding C-terminal part of human tau

Molecular Weight: 45-68kDa

Positive Control: Alzheimer's disease brain tissue

Cellular Localization: Cytoplasmic Ab-3 stains the human neurofibrillary tangles, neuropil threads, and neuritic plaques associated with Alzheimer's disease. It also stains astrocytes and ribosomes.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen TAU Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein encoding C-terminal part of human tau
Purification Method Alzheimer's disease brain tissue
Quantity 500 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.