Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1 Mouse Monoclonal Antibody

Catalog No. MS1105R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
Prediluted
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1105R1 1 mL Prediluted
MS1105S1 500 μL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MS1105R1 Supplier Epredia™ Supplier No. MS1105R1

TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.

Host Species: Mouse

Clone: SEN28

Isotype: IgG2a

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein corresponding to the amino terminal region of the TdT molecule

Molecular Weight: 58kDa

Positive Control: Thymus

Cellular Localization: Nuclear Ab-1 will be of use in the differentiation of lymphoblastic lymphoma from other lymphomas.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)


Specifications

Antigen TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SEN28
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to the amino terminal region of the TdT molecule
Quantity 1 mL
Regulatory Status IVD
Research Discipline DNA Repair
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C, Do Not Freeze
Form Prediluted
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.