Learn More
Epredia™ Lab Vision™ TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1 Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: SEN28
Isotype: IgG2a
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: N-terminal
Immunogen: Recombinant protein corresponding to the amino terminal region of the TdT molecule
Molecular Weight: 58kDa
Positive Control: Thymus
Cellular Localization: Nuclear Ab-1 will be of use in the differentiation of lymphoblastic lymphoma from other lymphomas.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SEN28 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to the amino terminal region of the TdT molecule |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Research Discipline | DNA Repair |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.