Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombospondin (TSP) Ab-4 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS421B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS421B1 500 μL Biotin Concentrated 200μg/mL
MS421B0 100 μL Biotin Concentrated 200μg/mL
MS421B 1 mL Biotin Concentrated 200μg/mL
MS421P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS421PABX 200 μL Unconjugated Concentrated 1mg/mL
MS421P1 500 μL Unconjugated Concentrated 200μg/mL
MS421P 1 mL Unconjugated Concentrated 200μg/mL
MS421R1 1 mL Unconjugated Prediluted Prediluted
MS421R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS421B1 Supplier Epredia Supplier No. MS421B1
Add to Cart
Add to Cart

Provide accurate, reproducible results Epredia™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: A6.1

Isotype: IgG1

Species Reactivity: Human, Cow, Pig, Horse, Dog, Sheep, Mouse and Rat. Others not known.

Epitope: Collagen Type V-binding domain of TSP

Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Positive Control: Tonsil or Placenta

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Thrombospondin (TSP) Ab-4
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A6.1
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Quantity 500 μL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Target Species Bovine, Canine, Equine, Human, Mouse, Pig, Rat, Sheep
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.