Learn More
Description
Host Species: Mouse
Clone: A6.1
Isotype: IgG1
Species Reactivity: Human, Cow, Pig, Horse, Dog, Sheep, Mouse and Rat. Others not known.
Epitope: Collagen Type V-binding domain of TSP
Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)
Positive Control: Tonsil or Placenta
Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
- Electron Microscopy
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Thrombospondin (TSP) Ab-4 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | A6.1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.