Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid Stimulating Hormone (TSH) Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1453R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1453R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1453R1 Supplier Epredia Supplier No. MS1453R1

Provide accurate, reproducible results with the Epredia™ Thyroid Stimulating Hormone (TSH) Ab-3, Mouse Monoclonal Antibody.

TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones.

Host Species: Mouse

Clone: TSH01 + TSH02

Isotype: IgG1/κ + IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human TSH-β

Molecular Weight: 14kDa (β), 28kDa (αβ)

Positive Control: Anterior pituitary

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)

Specifications

Antigen Thyroid Stimulating Hormone (TSH) Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TSH01 + TSH02
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Purified human TSH-Beta
Quantity 1 mL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.