Learn More
Description
Host Species: Mouse
Clone: 102D1
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human TIMP-1
Molecular Weight: 28.5kDa
Positive Control: Conditioned, serum-free medium from (TPA-treated) human fetal lung cell line HFL-1; Bladder, breast and ovarian CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | TIMP-1 Ab-2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 102D1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human TIMP-1 |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.