Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-1 Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS608R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS608R7 7 mL Unconjugated Prediluted Prediluted
MS608PABX 200 μL Unconjugated Concentrated 1mg/mL
MS608P1 500 μL Unconjugated Concentrated 200μg/mL
MS608P 1 mL Unconjugated Concentrated 200μg/mL
MS608B0 100 μL Biotin Concentrated 200μg/mL
MS608B1 500 μL Biotin Concentrated 200μg/mL
MS608B 1 mL Biotin Concentrated 200μg/mL
MS608P0 100 μL Unconjugated Concentrated 200μg/mL
MS608P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS608R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS608R7 Supplier Epredia Supplier No. MS608R7

Provide accurate, reproducible results with the Epredia™ TIMP-1 Ab-2, Mouse Monoclonal Antibody.

Tissue inhibitor of metalloproteinases-1 (TIMP-1) and Tissue inhibitor of metalloproteinases-2 (TIMP-2) are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-1 forms a complex of 1:1 stoichiometry with activated interstitial collagenase, activated stromelysin, active form of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A), and latent and active forms 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMPs inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.

Host Species: Mouse

Clone: 102D1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human TIMP-1

Molecular Weight: 28.5kDa

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fetal lung cell line HFL-1; Bladder, breast and ovarian CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen TIMP-1 Ab-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 102D1
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human TIMP-1
Quantity 7 mL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.