Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-2 Ab-5 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1485B0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS1485B0 100 μL Biotin Concentrated 200μg/mL
MS1485PABX 200 μL Unconjugated Concentrated 1mg/mL
MS1485P1 500 μL Unconjugated Concentrated 200μg/mL
MS1485B1 500 μL Biotin Concentrated 200μg/mL
MS1485B 1 mL Biotin Concentrated 200μg/mL
MS1485P0 100 μL Unconjugated Concentrated 200μg/mL
MS1485P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS1485P 1 mL Unconjugated Concentrated 200μg/mL
MS1485R1 1 mL Unconjugated Prediluted Prediluted
MS1485R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS1485B0 Supplier Epredia Supplier No. MS1485B0

Provide accurate, reproducible results with the Epredia™ TIMP-2 Ab-5, Mouse Monoclonal Antibody.

TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.

Host Species: Mouse

Clone: 3A4

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Rabbit, Cow and Guinea Ppig. Others not known.

Epitope: N-terminal

Immunogen: A synthetic peptide from the N-terminal region of human TIMP-2 protein

Molecular Weight: 21kDa

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells; Pancreatic or colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen TIMP-2 Ab-5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3A4
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen A synthetic peptide from the N-terminal region of human TIMP-2 protein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Bovine, Guinea Pig, Human, Mouse, Rabbit, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.