Learn More
Description
Host Species: Mouse
Clone: 3A4
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Rabbit, Cow and Guinea Ppig. Others not known.
Epitope: N-terminal
Immunogen: A synthetic peptide from the N-terminal region of human TIMP-2 protein
Molecular Weight: 21kDa
Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells; Pancreatic or colon CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
- Western Blotting
- Immunoprecipitation (Not suitable)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | TIMP-2 Ab-5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 3A4 |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A synthetic peptide from the N-terminal region of human TIMP-2 protein |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.