Description
Like the DNA chip, which scans the genome for alterations or abnormal gene expression, the Tissue Micro Array (TMA) permits parallel processing of biological samples. The TMA technology can be used to identify diagnostic markers, parameters to predict aggressiveness of cancer, and other biological characteristics of tumor cells.
Features:
- Mold is used to create a recipient paraffin block with 24 x 2.0mm holes arranged in a 6 x 4 array
- Tissue punch extractor takes cores from existing paraffin-embedded tissue blocks and injects the tissue cores into the holes of the recipient block
- Blocks can be sliced into hundreds of consecutive sections of 3 to 4μm each, making it possible to rapidly analyze hundreds of parameters in the same set of specimens
Includes:
- Two-piece mold set
- Punch extractor
- Mold screws
Recommended For:
- High throughput screening
- Rapid screening of novel gene or protein expressions
Specifications
Specifications
| Description | 6 x 4 array TMA Builder |
| Type | Tissue Microarray Builder |
| Array | 6 x 4 |