Learn More
Transforming Growth Factor α (TGFα) Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: MF9
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Rat and Mouse. Others not known.
Epitope: C-terminal
Immunogen: A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGFα
Molecular Weight: 6kDa
Positive Control: Recombinant TGFα protein, A2058 cells
Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with EGF and the neuropeptide synenkephalin. Ab-1 is excellent for staining of formalin-fixed, paraffin-embedded tissues. The staining with Ab-1 is completely blocked by the peptide used for raising this antibody.
Recommended for:
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Transforming Growth Factor alpha (TGFalpha) Ab-1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | MF9 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A 17-amino acid synthetic peptide from the carboxyl-terminus of Rat TGFalpha |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.