Learn More
Description
Host Species: Mouse
Clone: CUB 7402
Isotype: IgG1/κ
Species Reactivity: Human, Dog, Rabbit and Guinea Pig. Does not react with mouse and rat. Others not known.
Epitope: aa 447-478
Immunogen: Purified guinea pig liver TGase II
Molecular Weight: 77-85kDa
Positive Control: HUVEC cells, Breast CA
Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-1 inhibits the TGase II activity.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Affinity Purification
- Biological Blockade
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Transglutaminase II Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | CUB 7402 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Purified guinea pig liver TGase II |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.