Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transglutaminase II Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS224P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS224P1 500 μL Unconjugated Concentrated 200μg/mL
MS224P 1 mL Unconjugated Concentrated 200μg/mL
MS224B0 100 μL Biotin Concentrated 200μg/mL
MS224B1 500 μL Biotin Concentrated 200μg/mL
MS224B 1 mL Biotin Concentrated 200μg/mL
MS224P0 100 μL Unconjugated Concentrated 200μg/mL
MS224P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS224PABX 200 μL Unconjugated Concentrated 1mg/mL
MS224R1 1 mL Unconjugated Prediluted -
MS224R7 7 mL Unconjugated Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS224P1 Supplier Epredia Supplier No. MS224P1
Add to Cart
Add to Cart

Epredia™ Transglutaminase II Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Mouse

Clone: CUB 7402

Isotype: IgG1/κ

Species Reactivity: Human, Dog, Rabbit and Guinea Pig. Does not react with mouse and rat. Others not known.

Epitope: aa 447-478

Immunogen: Purified guinea pig liver TGase II

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-1 inhibits the TGase II activity.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Affinity Purification
  • Biological Blockade

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Transglutaminase II Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CUB 7402
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified guinea pig liver TGase II
Quantity 500 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Canine, Guinea Pig, Human, Rabbit
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.