Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Transglutaminase II Ab-2 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS279P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS279P0 100 μL Unconjugated Concentrated 200μg/mL
MS279B0 100 μL Biotin Concentrated 200μg/mL
MS279P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS279PABX 200 μL Unconjugated Concentrated 1mg/mL
MS279B1 500 μL Biotin Concentrated 200μg/mL
MS279P1 500 μL Unconjugated Concentrated 200μg/mL
MS279B 1 mL Biotin Concentrated 200μg/mL
MS279P 1 mL Unconjugated Concentrated 200μg/mL
MS279R1 1 mL Unconjugated Prediluted -
MS279R7 7 mL Unconjugated Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS279P0 Supplier Epredia™ Supplier No. MS279P0

Provides accurate, reproducible results with the Epredia™ Transglutaminase II Ab-2, Mouse Monoclonal Antibody.

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Mouse

Clone: TG100

Isotype: IgG1

Species Reactivity: Human, Rat and Guinea Pig. Others not known.

Epitope: aa 447-538

Immunogen: Purified guinea pig liver TGase II

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix Ab-2 is the Ab-of-Choice as a marker for endothelial cells in rat tissues

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Transglutaminase II Ab-2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TG100
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified guinea pig liver TGase II
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Guinea Pig, Human, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.