Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Transglutaminase II Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS300PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS300PABX 200 μL Concentrated
MS300P1ABX 100 μL Concentrated
MS300P 1 mL Concentrated
MS300R1 1 mL Prediluted
MS300R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS300PABX Supplier Epredia™ Supplier No. MS300PABX

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Provide accurate, reproducible results with the Epredia™ Transglutaminase II Ab-3, Mouse Monoclonal Antibody.

Transglutaminase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. It is also known as TGC, tTG, type II-, Gh, cytosolic-, liver-, endothelial-, erythrocyte-, cellular-transglutaminase. Different tissues and cell types express varying amounts of tissue transglutaminase with a markedly hugh expression in rheumatoid lesions. It is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

Host Species: Mouse

Clone: CUB 7402 + TG100

Isotype: IgG1

Species Reactivity: Human and Guinea Pig. Others not known.

Immunogen: Purified transglutaminase II from guinea pig liver

Molecular Weight: 77-85kDa

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasm, membrane and extracellular matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Transglutaminase II Ab-3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CUB 7402 + TG100
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified transglutaminase II from guinea pig liver
Quantity 200 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Guinea Pig, Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.