Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Tropomyosin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1256R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1256R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1256R1 Supplier Epredia™ Supplier No. MS1256R1

Provide accurate, reproducible results with the Epredia™ Tropomyosin Ab-1, Mouse Monoclonal Antibody.

Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.

Host Species: Mouse

Clone: CH1

Isotype: IgG1/κ

Species Reactivity: Human, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart and gizzard

Positive Control: Skeletal or Cardiac muscle

Cellular Localization: Cytoplasmic Ab-1 reacts strongly with striated muscle forms of tropomyosin, including cardiac alpha-tropomyosin and skeletal alpha- and Beta-tropomyosins. ^

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Tropomyosin Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CH1
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Tropomyosin protein purified from chicken embryo fibroblasts (CEF) and chicken leg, heart, and gizzard
Quantity 1 mL
Regulatory Status RUO
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Chicken, Human, Rat
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.