Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin T (Cardiac Isoform) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS295P Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
Prediluted
Concentration:
200μg/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS295P 1 mL Concentrated 200μg/mL
MS295P1 500 μL Concentrated 200μg/mL
MS295R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS295P Supplier Epredia Supplier No. MS295P
Add to Cart
Add to Cart

Epredia™ Troponin T, Cardiac Isoform Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Troponin T is a thin filament regulatory protein of striated muscle that binds to troponin C and I and to tropomyosin and is required for calcium-dependent ATPase activity of myofibrillar proteins. It is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. This expression in the human heart is affected by heart failure.

Host Species: Mouse

Clone: 13-11

Isotype: IgG1

Species Reactivity: Human, Pig, Dog, Rabbit, Rat, Mouse, Guinea Pig, Chicken, Sparrow and Goldfish. Does not react with frog.

Epitope: FMPNLVPPKI (5' region of the cardiac gene exon 10)

Immunogen: Purified rabbit cardiac troponin T isoform (TnT4R)

Positive Control: Cardiac muscle

Cellular Localization: Mostly over the I bands in myofibrils Ab-1 specifically recognizes the cardiac isoform of Troponin T (cTnT).

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Electron Microscopy

Specifications

Antigen Troponin T, Cardiac Isoform Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 13-11
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified rabbit cardiac troponin T isoform (TnT4R)
Quantity 1 mL
Regulatory Status IVD
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Goldfish, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sparrow
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.