Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTF-1 (Thyroid Transcription Factor-1) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS699P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Purified
Ready-to-Use
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form
MS699P0 100 μL Unconjugated Purified
MS699P 1 mL Unconjugated Concentrated
MS699P1 500 μL Unconjugated Purified
MS699RQ 12 mL Unconjugated Ready-to-Use
MS699B0 100 μL Biotin Concentrated
MS699B1 500 μL Biotin Concentrated
MS699B 1 mL Biotin Concentrated
MS699P1ABX 100 μL Unconjugated Concentrated
MS699PABX 200 μL Unconjugated Concentrated
MS699R1 1 mL Unconjugated Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS699P0 Supplier Epredia Supplier No. MS699P0

Provide accurate, reproducible results with the Epredia™ TTF-1 (Thyroid Transcription Factor-1) Ab-1, Mouse Monoclonal Antibody.

TTF-1(Thyroid transcription factor-1) is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. TTF-1 is detected in primary lung adenocarcinomas and small cell carcinomas and is absent in colon and breast carcinomas. Staining with TTF-1 antibody is useful for distinguishing between tumors of lung and non-lung origin.

Host Species: Mouse

Clone: 8G7G3/1

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Shows a broad species reactivity.

Epitope: Not determined

Immunogen: Rat TTF-1 recombinant protein

Molecular Weight: 40kDa

Positive Control: MAD109 cells, normal thyroid or lung

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Antigen TTF-1 (Thyroid Transcription Factor-1) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8G7G3/1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Rat TTF-1 recombinant protein
Quantity 100 μL
Regulatory Status IVD
Research Discipline Transcription Regulation
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.