Learn More
TTF-1 (Thyroid Transcription Factor-1) Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 8G7G3/1
Isotype: IgG1/κ
Species Reactivity: Human, Mouse and Rat. Shows a broad species reactivity.
Epitope: Not determined
Immunogen: Rat TTF-1 recombinant protein
Molecular Weight: 40kDa
Positive Control: MAD109 cells, normal thyroid or lung
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Specifications
| Antigen | TTF-1 (Thyroid Transcription Factor-1) Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 8G7G3/1 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Rat TTF-1 recombinant protein |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.