Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin-α Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS581P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS581P0 100 μL Concentrated 200μg/mL
MS581PABX 200 μL Concentrated 1mg/mL
MS581P1 500 μL Concentrated 200μg/mL
MS581P 1 mL Concentrated 200μg/mL
MS581P1ABX 100 μL Concentrated 1mg/mL
MS581R1 1 mL Prediluted Prediluted
MS581R7 7 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS581P0 Supplier Epredia Supplier No. MS581P0

Provide accurate, reproducible results with the Epredia™ Tubulin-α Ab-2, Mouse Monoclonal Antibody.

Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: α and Β. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.

Host Species: Mouse

Clone: DM1A

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Pig, Rat, Mouse, Guinea Pig, Gerbil, Frog and Chicken. Others not known.

Epitope: aa 426-450

Immunogen: Native chick brain microtubules

Molecular Weight: 57kDa

Positive Control: Ls174T, MAD109 cells; skin or lung

Cellular Localization: Cytoplasmic Ab-2 causes the 10nm filaments to collapse into large lateral aggregates collecting in the cell periphery or tight juxtanuclear caps. It does not block microtubule assembly. It does not inhibit polymerization or depolymerization of platelet tubulin in vitro. It blocks (by 70-80%) the ability of tubulin dimers (with GppNHp bound) to promote a stable inhibition of adenylyl cyclase.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Tubulin, alpha Ab-2
Applications Immunoassay, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DM1A
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Native chick brain microtubules
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Bovine, Chicken, Frog, Gerbil, Guinea Pig, Human, Mouse, Pig, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.