Learn More
Epredia™ Lab Vision™ Tubulin-Β Ab-6 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: TBN06; same as Tub 2.5
Isotype: IgG3
Species Reactivity: Human, Cow, Rabbit, Mouse, Chicken, Rat, Frog, Moth and Wheat. Others not known.
Epitope: Not determined
Immunogen: Purified tubulin
Molecular Weight: 55kDa
Positive Control: Ls174T, MAD109 cells; Skin or lung
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Tubulin-beta Ab-6 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | TBN06; same as Tub 2.5 |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Mouse |
| Immunogen | Purified tubulin |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.