Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vascular Endothelial Growth Factor (VEGF) Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS350P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
Concentration:
1mg/mL
200μg/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS350P1ABX 100 μL 1mg/mL
MS350P1 500 μL 200μg/mL
MS350PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS350P1ABX Supplier Epredia Supplier No. MS350P1ABX

Provide accurate, reproducible results with the Epredia™ Vascular Endothelial Growth Factor (VEGF) Ab-3, Mouse Monoclonal Antibody.

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: JH121

Isotype: IgG1/κ

Species Reactivity: Human and Rabbit. Others not known.

Epitope: Not determined

Immunogen: Recombinant human VEGF121

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein, Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-3 is excellent for neutralizing the bioactivity of human VEGF.

Recommended for:

  • Neutralizes Bioactivity of VEGF

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF) Ab-3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone JH121
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human VEGFsubscript121subscript
Quantity 100 μL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Target Species Human, Rabbit
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.