Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vascular Endothelial Growth Factor (VEGF) Ab-7 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1467R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1467R1 1 mL Prediluted -
MS1467P 1 mL Concentrated 200μg/mL
MS1467P1ABX 100 μL Concentrated 1mg/mL
MS1467PABX 200 μL Concentrated 1mg/mL
MS1467P1 500 μL Concentrated 200μg/mL
MS1467R7 7 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1467R1 Supplier Epredia Supplier No. MS1467R1

Provide accurate, reproducible results with the Epredia™ Vascular Endothelial Growth Factor (VEGF) Ab-7, Mouse Monoclonal Antibody.

VEGF which is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: VG1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant VEGF protein

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein, Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-7 does not neutralize the bioactivity of VEGF.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF) Ab-7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone VG1
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Recombinant VEGF protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.