Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Vascular Endothelial Growth Factor (VEGF) Rabbit Monoclonal Antibody

Catalog No. RM9128R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
7 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RM9128R7 7 mL
RM9128R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RM9128R7 Supplier Epredia™ Supplier No. RM9128R7

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Employ part of a comprehensive antibody portfolio with the Epredia™ Vascular Endothelial Growth Factor, Rabbit Monoclonal Antibody. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. This antibody reacts with the 165, 189 and 121 amino acid splice variants of VEGF of human, to a lesser extent, mouse and rat.

Host Species: Rabbit

Clone: SP28

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminal of human VEGF

Molecular Weight: 19-22kDa (reduced) or 38-44kDa (nonreduced)

Positive Control: Angiosarcoma, VEGF recombinant protein

Cellular Localization: Cytoplasmic, cell surface in endothelial cells This clone (SP28) is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF)
Applications Immunohistochemistry
Classification Monoclonal
Clone SP28
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide derived from N-terminal of human VEGF
Purification Method Angiosarcoma, VEGF recombinant protein
Quantity 7 mL
Primary or Secondary Primary
Target Species Human
Form Ready-to-Use
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.