Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Villin Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1499P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1499P1 500 μL Concentrated
MS1499P 1 mL Concentrated
MS1499P0 100 μL Concentrated
MS1499R1 1 mL Prediluted
MS1499RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1499P1 Supplier Epredia Supplier No. MS1499P1
Add to Cart
Add to Cart

Villin Ab-1, Mouse Monoclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Villin can cap, nucleate, sever and bundle actin in a Ca and phosphoinositide-regulated manner. It is associated with the microvillar actin core bundle of intestinal and renal brush border implicated in adsorption. Villin is composed of six repeats, each containing 150 residues that together constitute the core domain followed by the carboxyl-terminal headpiece domain of 87 residues. The core domain retains the Ca dependent capping nucleating and severing activity, whereas the headpiece domain contributes towards actin filament bundling and binding F-actin, independently of Ca

Host Species: Mouse

Clone: CWWB1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Immunogen: Human Villin protein

Molecular Weight: 92.5kDa

Positive Control: Small intestine and kidney

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Villin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CWWB1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human Villin protein
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.