Learn More
Description
Host Species: Mouse
Clone: CWWB1
Isotype: IgG1
Species Reactivity: Human. Others not known.
Immunogen: Human Villin protein
Molecular Weight: 92.5kDa
Positive Control: Small intestine and kidney
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Villin Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CWWB1 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human Villin protein |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Research Discipline | Cytoskeletal Protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.