Learn More
Description
Host Species: Mouse
Clone: V9
Isotype: IgG1/κ
Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.
Epitope: Not determined
Immunogen: Purified vimentin from pig eye lens
Molecular Weight: 57-60kDa
Positive Control: Sarcomas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Vimentin Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | V9 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Purified vimentin from pig eye lens |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.