Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS129P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS129P1 500 μL Unconjugated Concentrated 200μg/mL
MS129B0 100 μL Biotin Concentrated 200μg/mL
MS129PABX 200 μL Unconjugated Concentrated 1mg/mL
MS129P 1 mL Unconjugated Concentrated 200μg/mL
MS129RQ 12 mL Unconjugated Ready-to-Use Prediluted
MS129B1 500 μL Biotin Concentrated 200μg/mL
MS129B 1 mL Biotin Concentrated 200μg/mL
MS129P1ABX 100 μL Unconjugated Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS129P1 Supplier Epredia Supplier No. MS129P1
Add to Cart
Add to Cart

Epredia™ Vimentin Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.

Host Species: Mouse

Clone: V9

Isotype: IgG1/κ

Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.

Epitope: Not determined

Immunogen: Purified vimentin from pig eye lens

Molecular Weight: 57-60kDa

Positive Control: Sarcomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Vimentin Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone V9
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified vimentin from pig eye lens
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Equine, Feline, Gerbil, Hamster, Human, Mouse, Non-human Primate, Pig, Rabbit, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.