Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Vimentin Rabbit Monoclonal Antibody

Catalog No. RM9120S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9120S0 100 μL Concentrated
RM9120S1 500 μL Concentrated
RM9120S 1 mL Concentrated
RM9120R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RM9120S0 Supplier Epredia™ Supplier No. RM9120S0

Vimentin, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.

Host Species: Rabbit

Clone: SP20

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein encoding human vimentin

Molecular Weight: 57-60kDa

Positive Control: Sarcoma and melanoma

Cellular Localization: Cytoplasmic Clone SP20 is excellent for staining of formalin/paraffin with no special pretreatment.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Vimentin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP20
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein encoding human vimentin
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.