Learn More
Description
Host Species: Rabbit
Clone: SP20
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein encoding human vimentin
Molecular Weight: 57-60kDa
Positive Control: Sarcoma and melanoma
Cellular Localization: Cytoplasmic Clone SP20 is excellent for staining of formalin/paraffin with no special pretreatment.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Vimentin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP20 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encoding human vimentin |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Research Discipline | Cytoskeletal Protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.