Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Vinculin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1209P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1209P0 100 μL Concentrated 200μg/mL
MS1209P1 500 μL Concentrated 200μg/mL
MS1209P 1 mL Concentrated 200μg/mL
MS1209R7 7 mL Prediluted -
MS1209P1ABX 100 μL Concentrated 1mg/mL
MS1209PABX 200 μL Concentrated 1mg/mL
MS1209R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1209P0 Supplier Epredia™ Supplier No. MS1209P0

Provide accurate, reproducible results with the Epredia™ Vinculin Ab-1, Mouse Monoclonal Antibody.

Vinculin is an attachment protein involved in the indirect binding of intracellular actin filaments to extracellular fibronectin. Vinculin binds to actin capping protein and α actinin, which in turn binds to actin. The binding of fibronectin is also indirect where vinculin binds to talin, which in turn binds to the intracellular domain of the fibronectin receptor which itself is the ligand for fibronectin. Vinculin is widely distributed in tissue and is expressed where smooth muscle actin and fibroblasts attach to the extracellular matrix.

Host Species: Mouse

Clone: VLN01

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Semi-purified vinculin

Molecular Weight: 130kDa (vinculin) and 150kDa (meta-vinculin)

Positive Control: HeLa cells, skin

Cellular Localization: Cell membrane and cytoplasmic Ab-1 is useful for studies of vinculin in platelet activation.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Vinculin Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone VLN01
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Semi-purified vinculin
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.