Learn More
Epredia™ Lab Vision™ Vinculin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: VLN01
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Semi-purified vinculin
Molecular Weight: 130kDa (vinculin) and 150kDa (meta-vinculin)
Positive Control: HeLa cells, skin
Cellular Localization: Cell membrane and cytoplasmic Ab-1 is useful for studies of vinculin in platelet activation.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Vinculin Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | VLN01 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Semi-purified vinculin |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.