Learn More
Description
Host Species: Rat
Clone: 9A7γ.E10.E4
Isotype: IgG2b
Species Reactivity: Human, Pig, Mouse, Rat, Hamster, Frog, Chicken and Fish. Others not known.
Epitope: aa 89-105 (near the DNA-binding domain)
Immunogen: Vitamin D receptor protein from chicken intestine
Molecular Weight: 53kDa
Positive Control: LNCAP cells, HeLa cells
Cellular Localization: Nuclear Ab-1 reacts with both the occupied and unoccupied forms of VDR and shows no cross reaction with estrogen and glucocorticoid receptors. It is excellent for biochemical and histochemical applications.
Recommended for:
- Western Blotting
- Gel Supershift Assay
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Vitamin D Receptor (VDR) Ab-1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 9A7gamma.E10.E4 |
| Concentration | 200µg/mL |
| Conjugate | Biotin |
| Host Species | Rat |
| Immunogen | Vitamin D receptor protein from chicken intestine |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.