Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D Receptor (VDR) Ab-1 Rat Monoclonal Antibody, Epredia™

Catalog No. RT200P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200µg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
RT200P0 100 μL Unconjugated 200µg/mL
RT200P1 500 μL Unconjugated 200µg/mL
RT200B0 100 μL Biotin 200µg/mL
RT200B1 500 μL Biotin 200µg/mL
RT200B 1 mL Biotin 200µg/mL
RT200P 1 mL Unconjugated 200µg/mL
RT200P1ABX 100 μL Unconjugated 1mg/mL
RT200PABX 200 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. RT200P0 Supplier Epredia Supplier No. RT200P0
Add to Cart
Add to Cart

Provide accurate, reproducible results with the Epredia™ Vitamin D Receptor (VDR) Ab-1, Rat Monoclonal Antibody.

VDR is a member of super family of steroid hormone receptors. It is known to play role in cellular growth and differentiation, calcium and phosphorus metabolism and bone remodeling. Human breast carcinomas, leukemias, and prostatic carcinoma cells express VDR, which can modify their growth and differentiation. In avians, VDR appears as a doublet of 58kDa and 60kDa.

Host Species: Rat

Clone: 9A7γ.E10.E4

Isotype: IgG2b

Species Reactivity: Human, Pig, Mouse, Rat, Hamster, Frog, Chicken and Fish. Others not known.

Epitope: aa 89-105 (near the DNA-binding domain)

Immunogen: Vitamin D receptor protein from chicken intestine

Molecular Weight: 53kDa

Positive Control: LNCAP cells, HeLa cells

Cellular Localization: Nuclear Ab-1 reacts with both the occupied and unoccupied forms of VDR and shows no cross reaction with estrogen and glucocorticoid receptors. It is excellent for biochemical and histochemical applications.

Recommended for:

  • Western Blotting
  • Gel Supershift Assay

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Vitamin D Receptor (VDR) Ab-1
Applications Western Blot
Classification Monoclonal
Clone 9A7gamma.E10.E4
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rat
Immunogen Vitamin D receptor protein from chicken intestine
Quantity 100 μL
Regulatory Status RUO
Research Discipline Nuclear Receptors
Primary or Secondary Primary
Target Species Chicken, Fish, Frog, Hamster, Human, Mouse, Pig
Form Concentrated
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.