Learn More
Epredia™ Lab Vision™ Wilm's Tumor Protein (WT1) Rabbit Polyclonal Antibody, with BSA and azide
Epredia™ Wilm's Tumor Protein (WT1), Rabbit Polyclonal Antibody provides accurate, reproducible results.
Supplier: Epredia™ RB9267R1
Description
Wilm's Tumor (WT), a sporadic and familial childhood kidney tumor, is genetically heterogeneous. Wilm's tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed infetal kidney and mesothelioma, and its expression has been suggested as a marker for Wilm's tumor and mesothelioma.Host Species: Rabbit
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from the C-terminus of human Wilm's tumor protein
Molecular Weight: 47-55kDa
Positive Control: Wilm's tumor
Cellular Localization: Cytoplasmic and nuclear This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| Wilms Tumor Protein (WT1) | |
| Polyclonal | |
| Unconjugated | |
| A synthetic peptide derived from the C-terminus of human Wilms tumor protein | |
| RUO | |
| Primary | |
| Prediluted |
| Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| Rabbit | |
| 1 mL | |
| Cancer and Tumor Biology | |
| Human, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.